Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Muc30E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205954.5 | NP_995676.4 | 1 | 5059 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |