Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273191.1 | NP_001260120.1 | 4 - 5 | 911 | 65 | |
NM_135134.3 | NP_608978.2 | 4 - 5 | 917 | 65 | ||
GenBank mRNA | AY119076.1 | - | 4 - 5 | 881 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |