Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pez | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273198.1 | NP_001260127.1 | 3 - 4 | 2415 | 69 | |
NM_135169.3 | NP_609013.1 | 2 - 3 | 2324 | 69 | ||
GenBank mRNA | AF217284.1 | - | 2 - 3 | 2282 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |