Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oatp26F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135211.3 | NP_609055.2 | 4 - 5 | 926 | 81 | |
GenBank mRNA | AY075472.1 | - | 4 - 5 | 923 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |