Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oatp26F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135211.3 | NP_609055.2 | 5 - 6 | 1859 | 56 | |
GenBank mRNA | AY075472.1 | - | 5 - 6 | 1856 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |