Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppk11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001347761.1 | NP_001334672.1 | 7 - 8 | 3005 | 98 |
Gene Symbol: ppk16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001347762.1 | NP_001334673.1 | 7 - 8 | 3005 | 98 | |
GenBank mRNA | AY226546.1 | - | 4 - 5 | 1389 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |