Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201840.2 | NP_001188769.1 | 6 - 7 | 887 | 64 | |
NM_001201841.2 | NP_001188770.1 | 6 - 7 | 849 | 64 | ||
NM_001201842.2 | NP_001188771.1 | 6 - 7 | 815 | 64 | ||
NM_001298866.1 | NP_001285795.1 | 6 - 7 | 991 | 64 | ||
NM_001298867.1 | NP_001285796.1 | 6 - 7 | 812 | 64 | ||
NM_001298868.1 | NP_001285797.1 | 6 - 7 | 1805 | 64 | ||
NM_135513.5 | NP_609357.2 | 6 - 7 | 991 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |