Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATAd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273397.1 | NP_001260326.1 | 4 - 5 | 2214 | 111 | |
NM_001273398.1 | NP_001260327.1 | 4 - 5 | 2034 | 111 | ||
NM_135539.3 | NP_609383.2 | 4 - 5 | 2034 | 111 | ||
GenBank mRNA | BT133312.1 | - | 4 - 5 | 2026 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |