Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RluA-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201853.2 | NP_001188782.2 | 8 - 9 | 1589 | 108 | |
NM_001273423.1 | NP_001260352.1 | 7 - 8 | 1697 | 108 | ||
NM_135567.4 | NP_609411.1 | 7 - 8 | 1722 | 108 | ||
GenBank mRNA | AY058686.1 | - | 7 - 8 | 1674 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |