Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014480.4 | NP_001014480.4 | 3 | 662 | 78 | |
NM_001169474.3 | NP_001162945.3 | 3 | 1363 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |