Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AstCC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298901.1 | NP_001285830.1 | 1 - 2 | 420 | 109 | |
NM_135639.4 | NP_609483.2 | 1 - 2 | 420 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |