Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135643.4 | NP_609487.2 | 5 - 6 | 2523 | 101 | |
GenBank mRNA | AY128416.1 | - | 5 - 6 | 2519 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |