Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spz4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135660.3 | NP_609504.2 | 2 - 3 | 1748 | 76 | |
GenBank mRNA | BT100208.1 | - | 2 - 3 | 1712 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |