Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dif | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169527.2 | NP_001162998.1 | 4 - 5 | 1565 | 63 | |
NM_001169528.1 | NP_001162999.1 | 4 - 5 | 1325 | 63 | ||
NM_078865.4 | NP_523589.2 | 4 - 5 | 1565 | 63 | ||
NM_165216.3 | NP_724050.1 | 4 - 5 | 1213 | 63 | ||
GenBank mRNA | BT025231.1 | - | 4 - 5 | 1209 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |