Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ham | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259152.1 | NP_001246081.1 | 5 - 6 | 1352 | 66 | |
NM_001273614.1 | NP_001260543.1 | 5 - 6 | 1684 | 66 | ||
NM_001273615.1 | NP_001260544.1 | 2 - 3 | 317 | 66 | ||
NM_001273616.1 | NP_001260545.2 | 4 - 5 | 1221 | 66 | ||
NM_165262.3 | NP_724130.4 | 5 - 6 | 1353 | 66 | ||
GenBank mRNA | AY166585.1 | - | 4 - 5 | 703 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |