Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259156.1 | NP_001246085.1 | 5 - 6 | 1412 | 67 | |
NM_001259157.3 | NP_001246086.1 | 4 - 5 | 1123 | 67 | ||
NM_001259158.3 | NP_001246087.1 | 4 - 5 | 695 | 67 | ||
NM_001259159.1 | NP_001246088.1 | 5 - 6 | 2054 | 67 | ||
NM_001273630.2 | NP_001260559.1 | 4 - 5 | 1123 | 67 | ||
NM_057258.5 | NP_476606.1 | 4 - 5 | 973 | 67 | ||
NM_165277.3 | NP_724161.1 | 5 - 6 | 1088 | 67 | ||
GenBank mRNA | BT029915.1 | - | 5 - 6 | 1085 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |