Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259156.1 | NP_001246085.1 | 6 - 7 | 1561 | 61 | |
NM_001259157.3 | NP_001246086.1 | 5 - 6 | 1272 | 61 | ||
NM_001259158.3 | NP_001246087.1 | 5 - 6 | 844 | 61 | ||
NM_001259159.1 | NP_001246088.1 | 6 - 7 | 2203 | 61 | ||
NM_001273630.2 | NP_001260559.1 | 5 - 6 | 1272 | 61 | ||
NM_057258.5 | NP_476606.1 | 5 - 6 | 1122 | 61 | ||
NM_165277.3 | NP_724161.1 | 6 - 7 | 1237 | 61 | ||
GenBank mRNA | BT029915.1 | - | 6 - 7 | 1234 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |