Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: swm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273657.1 | NP_001260586.1 | 5 - 6 | 3327 | 95 | |
NM_001273658.1 | NP_001260587.1 | 4 - 5 | 3293 | 95 | ||
NM_136132.5 | NP_609976.1 | 5 - 6 | 3324 | 95 | ||
NM_165304.2 | NP_724201.1 | 4 - 5 | 3290 | 95 | ||
GenBank mRNA | AY095056.1 | - | 5 - 6 | 3311 | 95 | |
EF601876.1 | - | 5 - 6 | 3313 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |