Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_165319.3 | NP_724244.1 | 3 - 4 | 520 | 63 | |
GenBank mRNA | AY061078.1 | - | 3 - 4 | 491 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |