Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spir | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_165325.3 | NP_724256.2 | 1 - 2 | 632 | 57 | |
GenBank mRNA | BT044175.1 | - | 1 - 2 | 607 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |