Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Src42A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057501.4 | NP_476849.1 | 8 - 9 | 1752 | 64 | |
NM_136347.2 | NP_610191.2 | 8 - 9 | 4838 | 64 | ||
GenBank mRNA | AY058652.1 | - | 8 - 9 | 1716 | 64 | |
BT044565.1 | - | 9 - 10 | 1620 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |