Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epac | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103732.2 | NP_001097202.2 | 15 - 16 | 2634 | 67 | |
NM_001273807.1 | NP_001260736.1 | 12 - 13 | 2278 | 67 | ||
NM_001273808.1 | NP_001260737.2 | 11 - 12 | 1897 | 67 | ||
NM_165490.2 | NP_724498.1 | 12 - 13 | 2279 | 67 | ||
GenBank mRNA | AY058255.1 | - | 12 - 13 | 2262 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |