Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: boca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_165541.2 | NP_724578.1 | 1 - 2 | 221 | 131 | |
GenBank mRNA | BT044294.1 | - | 1 - 2 | 218 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |