Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_165543.2 | NP_724582.1 | 9 - 10 | 3030 | 81 | |
NM_165544.2 | NP_724583.1 | 10 - 11 | 2866 | 81 | ||
GenBank mRNA | AY069520.1 | - | 9 - 10 | 2886 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |