Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103749.2 | NP_001097219.1 | 12 - 13 | 2032 | 81 | |
NM_001103750.3 | NP_001097220.1 | 11 - 12 | 1723 | 81 | ||
NM_001273863.1 | NP_001260792.1 | 11 - 12 | 1723 | 81 | ||
NM_001316408.1 | NP_001303337.1 | 11 - 12 | 1723 | 81 | ||
NM_165568.3 | NP_724619.2 | 13 - 14 | 2113 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |