Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup44A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136499.3 | NP_610343.1 | 1 - 2 | 199 | 77 | |
NM_165582.2 | NP_724637.1 | 2 - 3 | 274 | 77 | ||
NM_165583.2 | NP_724638.1 | 2 - 3 | 322 | 77 | ||
GenBank mRNA | AY060480.1 | - | 1 - 2 | 1403 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |