Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057547.5 | NP_476895.1 | 6 - 7 | 1208 | 80 | |
NM_165694.3 | NP_724822.1 | 6 - 7 | 1333 | 80 | ||
NM_165695.2 | NP_724823.2 | 6 - 7 | 1195 | 80 | ||
GenBank mRNA | BT099607.1 | - | 6 - 7 | 1179 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |