Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nub | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103683.2 | NP_001097153.1 | 3 | 1978 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |