Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RanBPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038849.2 | NP_001033938.1 | 2 - 3 | 1455 | 61 | |
NM_001259326.1 | NP_001246255.1 | 2 - 3 | 1455 | 61 | ||
NM_001273938.1 | NP_001260867.1 | 2 - 3 | 1106 | 61 | ||
NM_001316413.1 | NP_001303342.1 | 2 - 3 | 1459 | 61 | ||
NM_136741.3 | NP_610585.2 | 2 - 3 | 1109 | 61 | ||
NM_165770.3 | NP_724932.1 | 2 - 3 | 1459 | 61 | ||
GenBank mRNA | AY973545.1 | - | 2 - 3 | 1416 | 61 | |
AY973546.1 | - | 2 - 3 | 1416 | 61 | ||
BT150441.1 | - | 2 - 3 | 1109 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |