Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136821.3 | NP_610665.2 | 5 - 6 | 2145 | 82 | |
NM_165815.2 | NP_725019.1 | 5 - 6 | 1820 | 82 | ||
NM_165816.2 | NP_725020.1 | 5 - 6 | 1543 | 82 | ||
GenBank mRNA | BT023808.1 | - | 5 - 6 | 1796 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |