Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nullo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078512.5 | NP_511067.3 | 1 | 699 | 64 | |
GenBank mRNA | AY094877.1 | - | 1 | 697 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |