Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: org-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298090.1 | NP_001285019.1 | 6 - 7 | 1949 | 71 | |
NM_078530.3 | NP_511085.2 | 6 - 7 | 1749 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |