Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gr8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078542.6 | NP_511097.3 | 3 - 4 | 1247 | 89 | |
GenBank mRNA | BT024439.1 | - | 3 - 4 | 1247 | 89 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Drosophila melanogaster |