Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298141.1 | NP_001285070.1 | 1 - 2 | 285 | 60 | |
NM_078547.3 | NP_511102.3 | 1 - 2 | 287 | 60 | ||
GenBank mRNA | AY061042.1 | - | 1 - 2 | 267 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |