Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258680.2 | NP_001245609.1 | 10 - 11 | 2189 | 64 | |
NM_001258681.2 | NP_001245610.1 | 7 - 8 | 1931 | 64 | ||
NM_001258682.2 | NP_001245611.1 | 9 - 10 | 2058 | 64 | ||
NM_001272470.2 | NP_001259399.1 | 4 - 5 | 1694 | 64 | ||
NM_001272471.2 | NP_001259400.1 | 4 - 5 | 1691 | 64 | ||
NM_001272472.2 | NP_001259401.1 | 4 - 5 | 1691 | 64 | ||
NM_078549.4 | NP_511104.3 | 4 - 5 | 1691 | 64 | ||
NM_167222.3 | NP_727405.1 | 9 - 10 | 1676 | 64 | ||
GenBank mRNA | AY060359.1 | - | 4 - 5 | 820 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |