Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Or9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078552.3 | NP_511107.1 | 2 - 3 | 599 | 79 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Recommended Assay Drosophila melanogaster |