Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bif | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078573.4 | NP_511128.2 | 5 - 6 | 3638 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |