Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PhKgamma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298204.1 | NP_001285133.1 | 6 - 7 | 958 | 61 | |
NM_001298205.1 | NP_001285134.1 | 6 - 7 | 1164 | 61 | ||
NM_078574.4 | NP_511129.3 | 6 - 7 | 942 | 61 | ||
NM_167296.3 | NP_727548.1 | 6 - 7 | 1063 | 61 | ||
NM_167297.3 | NP_727549.2 | 6 - 7 | 1330 | 61 | ||
GenBank mRNA | AY051575.1 | - | 6 - 7 | 945 | 61 | |
AY051635.1 | - | 6 - 7 | 1069 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |