Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tsg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078580.4 | NP_511135.1 | 1 | 695 | 57 | |
GenBank mRNA | BT120366.1 | - | 1 | 659 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |