Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103491.1 | NP_001096961.1 | 4 - 5 | 1041 | 93 | |
NM_001298217.1 | NP_001285146.1 | 5 - 6 | 1421 | 93 | ||
NM_078581.3 | NP_511136.2 | 6 - 7 | 1801 | 93 | ||
GenBank mRNA | BT003214.1 | - | 6 - 7 | 1798 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |