Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ten-a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144717.4 | NP_001138189.1 | 15 - 16 | 5056 | 83 | |
NM_001144718.3 | NP_001138190.1 | 16 - 17 | 5761 | 83 | ||
NM_001169258.2 | NP_001162729.1 | 10 - 11 | 2990 | 83 | ||
NM_001169259.3 | NP_001162730.2 | 16 - 17 | 5166 | 83 | ||
NM_001169260.3 | NP_001162731.1 | 17 - 18 | 6032 | 83 | ||
NM_001169261.3 | NP_001162732.2 | 16 - 17 | 5083 | 83 | ||
NM_001258711.2 | NP_001245640.1 | 13 - 14 | 4005 | 83 | ||
NM_001258712.2 | NP_001245641.1 | 14 - 15 | 4038 | 83 | ||
NM_001258713.2 | NP_001245642.1 | 14 - 15 | 4050 | 83 | ||
NM_001272554.2 | NP_001259483.1 | 13 - 14 | 3905 | 83 | ||
NM_078582.4 | NP_511137.3 | 11 - 12 | 3191 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |