Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ten-a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144717.4 | NP_001138189.1 | 9 - 10 | 4027 | 60 | |
NM_001144718.3 | NP_001138190.1 | 10 - 11 | 4732 | 60 | ||
NM_001169258.2 | NP_001162729.1 | 4 - 5 | 1961 | 60 | ||
NM_001169259.3 | NP_001162730.2 | 10 - 11 | 4137 | 60 | ||
NM_001169260.3 | NP_001162731.1 | 11 - 12 | 5003 | 60 | ||
NM_001169261.3 | NP_001162732.2 | 9 - 10 | 4027 | 60 | ||
NM_001258711.2 | NP_001245640.1 | 7 - 8 | 2976 | 60 | ||
NM_001258712.2 | NP_001245641.1 | 7 - 8 | 2982 | 60 | ||
NM_001258713.2 | NP_001245642.1 | 7 - 8 | 2994 | 60 | ||
NM_001272554.2 | NP_001259483.1 | 7 - 8 | 2876 | 60 | ||
NM_078582.4 | NP_511137.3 | 5 - 6 | 2162 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |