Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ten-a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144717.4 | NP_001138189.1 | 13 - 14 | 4778 | 55 | |
NM_001144718.3 | NP_001138190.1 | 14 - 15 | 5483 | 55 | ||
NM_001169258.2 | NP_001162729.1 | 8 - 9 | 2712 | 55 | ||
NM_001169259.3 | NP_001162730.2 | 14 - 15 | 4888 | 55 | ||
NM_001169260.3 | NP_001162731.1 | 15 - 16 | 5754 | 55 | ||
NM_001258711.2 | NP_001245640.1 | 11 - 12 | 3727 | 55 | ||
NM_001272554.2 | NP_001259483.1 | 11 - 12 | 3627 | 55 | ||
NM_078582.4 | NP_511137.3 | 9 - 10 | 2913 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |