Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272571.2 | NP_001259500.1 | 2 - 3 | 1564 | 79 | |
NM_001272572.2 | NP_001259501.1 | 2 - 3 | 1564 | 79 | ||
NM_001298253.1 | NP_001285182.1 | 2 - 3 | 1564 | 79 | ||
NM_001382110.1 | NP_001368981.1 | 2 - 3 | 1564 | 79 | ||
NM_001382111.1 | NP_001368982.1 | 2 - 3 | 1564 | 79 | ||
NM_078587.3 | NP_511142.2 | 2 - 3 | 1564 | 79 | ||
NM_167346.2 | NP_727661.1 | 2 - 3 | 1564 | 79 | ||
GenBank mRNA | AY069695.1 | - | 2 - 3 | 1499 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |