Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rdgB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014740.2 | NP_001014740.1 | 4 - 5 | 784 | 63 | |
NM_001014741.3 | NP_001014741.1 | 5 - 6 | 842 | 63 | ||
NM_001169277.2 | NP_001162748.1 | 4 - 5 | 784 | 63 | ||
NM_001169278.3 | NP_001162749.1 | 4 - 5 | 784 | 63 | ||
NM_001169279.3 | NP_001162750.1 | 4 - 5 | 784 | 63 | ||
NM_001169280.3 | NP_001162751.1 | 4 - 5 | 784 | 63 | ||
NM_001272605.2 | NP_001259534.1 | 5 - 6 | 865 | 63 | ||
NM_001272606.1 | NP_001259535.1 | 4 - 5 | 899 | 63 | ||
NM_001298300.1 | NP_001285229.1 | 4 - 5 | 784 | 63 | ||
NM_078594.3 | NP_511149.2 | 3 - 4 | 828 | 63 | ||
NM_167382.3 | NP_727733.1 | 4 - 5 | 822 | 63 | ||
GenBank mRNA | AY051422.1 | - | 4 - 5 | 804 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |