Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NetB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169282.1 | NP_001162753.1 | 4 - 5 | 2209 | 89 | |
NM_001169283.2 | NP_001162754.1 | 5 - 6 | 2212 | 89 | ||
NM_001169284.2 | NP_001162755.1 | 5 - 6 | 2149 | 89 | ||
NM_001258739.2 | NP_001245668.1 | 5 - 6 | 2215 | 89 | ||
NM_001258740.2 | NP_001245669.1 | 5 - 6 | 2152 | 89 | ||
NM_001272617.1 | NP_001259546.1 | 4 - 5 | 2188 | 89 | ||
NM_078600.4 | NP_511155.1 | 5 - 6 | 2302 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |