Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272623.2 | NP_001259552.1 | 4 - 5 | 519 | 69 | |
NM_001272624.2 | NP_001259553.1 | 5 - 6 | 558 | 69 | ||
NM_001272625.2 | NP_001259554.1 | 4 - 5 | 489 | 69 | ||
NM_001272626.2 | NP_001259555.1 | 5 - 6 | 869 | 69 | ||
NM_078602.4 | NP_511157.2 | 4 - 5 | 489 | 69 | ||
NM_167415.3 | NP_727797.1 | 5 - 6 | 528 | 69 | ||
NM_167416.3 | NP_727798.1 | 5 - 6 | 640 | 69 | ||
NM_167417.3 | NP_727799.1 | 4 - 5 | 601 | 69 | ||
NM_167419.2 | NP_727812.2 | 1 - 2 | 881 | 69 | ||
GenBank mRNA | BT009962.1 | - | 4 - 5 | 487 | 69 | |
BT126133.1 | - | 5 - 6 | 507 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |