Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078603.4 | NP_511158.2 | 16 - 17 | 3261 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster Inventoried |