Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hiw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298329.1 | NP_001285258.1 | 11 - 12 | 8451 | 79 | |
NM_078604.4 | NP_511159.3 | 11 - 12 | 8451 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |