Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hiw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298329.1 | NP_001285258.1 | 7 - 8 | 4422 | 60 | |
NM_078604.4 | NP_511159.3 | 7 - 8 | 4422 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |